Butyl benzoate, 98+%
Butyl benzoate, 98+%
Butyl benzoate, 98+%
Thermo Scientific Chemicals

Butyl benzoate, 98+%

CAS: 136-60-7 | C11H14O2 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 296400010
Price (EUR)
99,70
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
99,70
Each
Chemical Identifiers
CAS136-60-7
IUPAC Namebutyl benzoate
Molecular FormulaC11H14O2
InChI KeyXSIFPSYPOVKYCO-UHFFFAOYSA-N
SMILESCCCCOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.0 %
Appearance (Form)Liquid
Refractive index1.4960 to 1.4990 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only