4-(4-Chlorophenyl)-4-hydroxypiperidine, 99+%, crystalline
4-(4-Chlorophenyl)-4-hydroxypiperidine, 99+%, crystalline
4-(4-Chlorophenyl)-4-hydroxypiperidine, 99+%, crystalline
Thermo Scientific Chemicals

4-(4-Chlorophenyl)-4-hydroxypiperidine, 99+%, crystalline

CAS: 39512-49-7 | C11H15Cl2NO | 248.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 296840010
Price (EUR)
27,10
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
27,10
Each
Chemical Identifiers
CAS39512-49-7
IUPAC Namehydrogen 4-(4-chlorophenyl)piperidin-4-ol chloride
Molecular FormulaC11H15Cl2NO
InChI KeyPXAJMBHRLGKPQV-UHFFFAOYSA-N
SMILES[H+].[Cl-].OC1(CCNCC1)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (1 g, 105°C, 2 h, 760 mmHg)
Sulfated ash=<0.2 %
UV(1 % w/v in DMF) 1 cm silica cell
Appearance (Color)White to cream
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only