6-Chloro-2-benzoxazolethiol, 99%, Thermo Scientific Chemicals
6-Chloro-2-benzoxazolethiol, 99%, Thermo Scientific Chemicals
6-Chloro-2-benzoxazolethiol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Chloro-2-benzoxazolethiol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 297300050
Price (EUR)
24,20
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
24,20
Each
Chemical Identifiers
CAS22876-20-6
IUPAC Name6-chloro-2,3-dihydro-1,3-benzoxazole-2-thione
Molecular FormulaC7H4ClNOS
InChI KeyHAASPZUBSZGCKU-UHFFFAOYSA-N
SMILESClC1=CC=C2NC(=S)OC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Melting point229°C to 232°C
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only