3-Bromo-1,1,1-trifluoroacetone, 97%
3-Bromo-1,1,1-trifluoroacetone, 97%
3-Bromo-1,1,1-trifluoroacetone, 97%
3-Bromo-1,1,1-trifluoroacetone, 97%
Thermo Scientific Chemicals

3-Bromo-1,1,1-trifluoroacetone, 97%

CAS: 431-35-6 | C3H2BrF3O | 190.95 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2974400505 gGlass bottle
Catalog number 297440050
Price (EUR)
49,50
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
49,50
Each
Add to cart
Chemical Identifiers
CAS431-35-6
IUPAC Name3-bromo-1,1,1-trifluoropropan-2-one
Molecular FormulaC3H2BrF3O
InChI KeyONZQYZKCUHFORE-UHFFFAOYSA-N
SMILESFC(F)(F)C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.3740 to 1.3770 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only