6-Methoxy-2-tetralone, 90%
6-Methoxy-2-tetralone, 90%
6-Methoxy-2-tetralone, 90%
Thermo Scientific Chemicals

6-Methoxy-2-tetralone, 90%

CAS: 2472-22-2 | C11H12O2 | 176.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 298920010
Price (EUR)
118,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
118,00
Each
Chemical Identifiers
CAS2472-22-2
IUPAC Name6-methoxy-1,2,3,4-tetrahydronaphthalen-2-one
Molecular FormulaC11H12O2
InChI KeyRMRKDYNVZWKAFP-UHFFFAOYSA-N
SMILESCOC1=CC=C2CC(=O)CCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Yellow to light beige to orange
GC>=88 %
Appearance (Form)Low melting crystalline solid
Melting point28°C to 35°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only