5-Nitrosalicylic acid, 99%
5-Nitrosalicylic acid, 99%
5-Nitrosalicylic acid, 99%
Thermo Scientific Chemicals

5-Nitrosalicylic acid, 99%

CAS: 96-97-9 | C7H5NO5 | 183.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
250 g
Packaging:
View all
Plastic bottle
Catalog number 300480250
Price (EUR)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS96-97-9
IUPAC Name2-hydroxy-5-nitrobenzoic acid
Molecular FormulaC7H5NO5
InChI KeyPPDRLQLKHRZIJC-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC=C1O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow to beige to orange to tan
HPLC>=98.5 %
Appearance (Form)Powder
Infrared spectrumConforms
Melting point228°C to 233°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only