Vinylphosphonic acid, 90%, technical, Thermo Scientific Chemicals
Vinylphosphonic acid, 90%, technical, Thermo Scientific Chemicals
Vinylphosphonic acid, 90%, technical, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vinylphosphonic acid, 90%, technical, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 300592500
Price (EUR)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS1746-03-8
IUPAC Nameethenylphosphonate
Molecular FormulaC2H3O3P
InChI KeyZTWTYVWXUKTLCP-UHFFFAOYSA-L
SMILES[O-]P([O-])(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration Acid>=88 %
Water=<1.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only