Diphenyl ditelluride, 97%
Diphenyl ditelluride, 97%
Diphenyl ditelluride, 97%
Diphenyl ditelluride, 97%
Thermo Scientific Chemicals

Diphenyl ditelluride, 97%

CAS: 32294-60-3 | C12H10Te2 | 409.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 301840050
Price (EUR)
153,00
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
153,00
Each
Add to cart
Chemical Identifiers
CAS32294-60-3
IUPAC Name(phenylditellanyl)benzene
Molecular FormulaC12H10Te2
InChI KeyVRLFOXMNTSYGMX-UHFFFAOYSA-N
SMILES[Te]([Te]C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point64°C to 67°C
Appearance (Form)Powder or crystals
Appearance (Color)Orange to brown
HPLC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only