Sulfuric acid, pure, 96.00% (+/- 0.50%) solution in water
Sulfuric acid, pure, 96.00% (+/- 0.50%) solution in water
Sulfuric acid, pure, 96.00% (+/- 0.50%) solution in water
Thermo Scientific Chemicals

Sulfuric acid, pure, 96.00% (+/- 0.50%) solution in water

CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Plastic bottle
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 302070026
Price (EUR)
97,90
Each
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
97,90
Each
Chemical Identifiers
CAS7664-93-9
IUPAC Namesulfuric acid
Molecular FormulaH2O4S
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N
SMILESOS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH95.50 to 96.50 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only