beta-Alanine ethyl ester hydrochloride, 98%
beta-Alanine ethyl ester hydrochloride, 98%
beta-Alanine ethyl ester hydrochloride, 98%
Thermo Scientific Chemicals

beta-Alanine ethyl ester hydrochloride, 98%

CAS: 4244-84-2 | C5H12ClNO2 | 153.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass Bottle
Catalog number 303060100
Price (EUR)
61,30
Each
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
61,30
Each
Chemical Identifiers
CAS4244-84-2
IUPAC Nameethyl 3-aminopropanoate hydrochloride
Molecular FormulaC5H12ClNO2
InChI KeyRJCGNNHKSNIUAT-UHFFFAOYSA-N
SMILESCl.CCOC(=O)CCN
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric97.5 to 102.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder
TLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only