Product Image
Thermo Scientific Chemicals

O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 99

CAS: 125700-67-6 | C1;1;H1;6;N5;O·BF4; | 321.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3031800505 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 303180050
Price (EUR)
37,60
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
37,60
Each
Chemical Identifiers
CAS125700-67-6
IUPAC Name[(1H-1,2,3-benzotriazol-1-yloxy)(dimethylamino)methylidene]dimethylazanium; tetrafluoroboranuide
Molecular FormulaC11H16BF4N5O
InChI KeyJKEKMBGUVUKMQB-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C12)=[N+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Loss on drying=<1 %
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only