3-Bromothiophenol, 95%
3-Bromothiophenol, 95%
3-Bromothiophenol, 95%
Thermo Scientific Chemicals

3-Bromothiophenol, 95%

CAS: 6320-01-0 | C6H5BrS | 189.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 303710010
Price (EUR)
39,10
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
39,10
Each
Chemical Identifiers
CAS6320-01-0
IUPAC Name3-bromobenzene-1-thiol
Molecular FormulaC6H5BrS
InChI KeyHNGQQUDFJDROPY-UHFFFAOYSA-N
SMILESSC1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=94 %
Refractive index1.6320 to 1.6350 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only