1-Bromopinacolone, 93%
1-Bromopinacolone, 93%
1-Bromopinacolone, 93%
Thermo Scientific Chemicals

1-Bromopinacolone, 93%

CAS: 5469-26-1 | C6H11BrO | 179.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 306410250
Price (EUR)
172,00
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
172,00
Each
Chemical Identifiers
CAS5469-26-1
IUPAC Name1-bromo-3,3-dimethylbutan-2-one
Molecular FormulaC6H11BrO
InChI KeySAIRZMWXVJEBMO-UHFFFAOYSA-N
SMILESCC(C)(C)C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to yellow-brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=91 %
Refractive index1.4656 to 1.4676 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only