alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene, 98%
alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene, 98%
alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene, 98%
Thermo Scientific Chemicals

alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene, 98%

CAS: 402-49-3 | C8H6BrF3 | 239.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 312990050
Price (EUR)
63,60
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
63,60
Each
Chemical Identifiers
CAS402-49-3
IUPAC Name1-(bromomethyl)-4-(trifluoromethyl)benzene
Molecular FormulaC8H6BrF3
InChI KeyIKSNDOVDVVPSMA-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)White to light yellow
Appearance (Form)Low melting crystalline solid
Infrared spectrumConforms
Melting point30°C to 34°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only