N,N-Diisopropylcarbamoyl chloride, 98%
N,N-Diisopropylcarbamoyl chloride, 98%
N,N-Diisopropylcarbamoyl chloride, 98%
Thermo Scientific Chemicals

N,N-Diisopropylcarbamoyl chloride, 98%

CAS: 19009-39-3 | C7H14ClNO | 163.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 314520250
Price (EUR)
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS19009-39-3
IUPAC NameN,N-bis(propan-2-yl)carbamoyl chloride
Molecular FormulaC7H14ClNO
InChI KeyRSAFAYLZKCYUQW-UHFFFAOYSA-N
SMILESCC(C)N(C(C)C)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or low melting solid
Infrared spectrumConforms
Appearance (Color)Colorless to white to light yellow
Melting point55°C to 60°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only