4-Methoxyphenyl chloroformate, 98%
4-Methoxyphenyl chloroformate, 98%
4-Methoxyphenyl chloroformate, 98%
Thermo Scientific Chemicals

4-Methoxyphenyl chloroformate, 98%

CAS: 7693-41-6 | C8H7ClO3 | 186.591 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 314570250
Price (EUR)
223,00
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
223,00
Each
Chemical Identifiers
CAS7693-41-6
IUPAC Name4-methoxyphenyl carbonochloridate
Molecular FormulaC8H7ClO3
InChI KeyCCFSGQKTSBIIHG-UHFFFAOYSA-N
SMILESCOC1=CC=C(OC(Cl)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.5220 to 1.5260 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only