4'-Chloro-2,2':6',2″-terpyridine, 99%
4'-Chloro-2,2':6',2″-terpyridine, 99%
4'-Chloro-2,2':6',2″-terpyridine, 99%
Thermo Scientific Chemicals

4'-Chloro-2,2':6',2″-terpyridine, 99%

CAS: 128143-89-5 | C15H10ClN3 | 267.72 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3196700101 gGlass bottle
Catalog number 319670010
Price (EUR)
169,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
169,00
Each
Chemical Identifiers
CAS128143-89-5
IUPAC Name4-chloro-6-(pyridin-2-yl)-2,2'-bipyridine
Molecular FormulaC15H10ClN3
InChI KeyAHEMFMCEBIJRMU-UHFFFAOYSA-N
SMILESClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey to green to brown
GC>=98.5 %
Appearance (Form)Crystalline powder or needles
Melting point149°C to 153°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only