Pierce™ ECL Western Blotting Substrate
Thermo Scientific™

Pierce™ ECL Western Blotting Substrate

Value-priced, entry-level peroxidase substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
32106X44 x 500 mL
3210950 mL
32209X44 x 250 mL
32209250 mL
32106500 mL
Catalog number 32106X4
Price (EUR)
1.018,65
Precio exclusivo compra web
1.132,00
Save 113,35 (10%)
Each
Quantity:
4 x 500 mL
Price (EUR)
1.018,65
Precio exclusivo compra web
1.132,00
Save 113,35 (10%)
Each

Thermo Scientific Pierce ECL Western Blotting Substrate is a value-priced, entry-level horseradish peroxidase (HRP) substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.

Pierce ECL Plus Western Blotting Substrate characteristics include:

  • Sensitivity: low picogram
  • Stability: 1-hr working solution stability, 1-year kit stability at 4°C
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: 1 to 2 hours
  • Recommended primary antibody concentration: 1:1,000–1:5,000 dilution (0.2–10 μg/mL)
  • Recommended secondary antibody concentration: 1:1,000–1:15,000 dilution (0.07–1.0 μg/mL)

Pierce ECL Western Blotting Substrate provides reliability and performance equivalent to other standard ECL substrates for detection of HRP enzyme activity. It enables the detection of picogram amounts of antigen and allows for easy detection of HRP using photographic or other imaging methods.

Because the luminol and peroxide reagent formulations are identical to other commercially available substrate products, one can switch to Pierce ECL without needing to optimize probing conditions or incubation protocols. Pierce ECL Western Blotting Substrate provides performance identical to the original Amersham ECL Western Blotting Detection Reagent from GE Healthcare.

For Research Use Only. Not for use in diagnostic procedures.

General References:

  1. Aupperlee, M.,et al.(2005).Endocrinology 146,3577-3588.
  2. Li, Y.,et al.(2004).Blood.104,1137-1144.
  3. MacMillan, D.,et al.(2005).J. Cell Sci.118,5443-5451.
Specifications
ConjugateHRP
DescriptionPierce ECL Western Blotting Substrate
Recommended Antibody Concentrations2°: 1:1-15 K (0.07-1 μg/mL),, 1°: 1:1 K (0.2-10 μg/mL)
SensitivityLow picogram
Signal Duration1 to 2 hr.
Detection MethodChemiluminescence
FormLiquid
Product LinePierce
Product TypeSubstrate
Quantity4 x 500 mL
SubstrateHorseradish Peroxidase
Unit SizeEach
Contents & Storage
Luminol/Enhancer, 4 x 250 mL; Stable Peroxide Buffer, 4 x 250 mL. Upon receipt store at 2-8°C. Product is shipped at ambient temperature.