trans-2-Pentenal, 97%
trans-2-Pentenal, 97%
trans-2-Pentenal, 97%
Thermo Scientific Chemicals

trans-2-Pentenal, 97%

CAS: 1576-87-0 | C5H8O | 84.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 325420050
Price (EUR)
37,20
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
37,20
Each
Chemical Identifiers
CAS1576-87-0
IUPAC Name(2E)-pent-2-enal
Molecular FormulaC5H8O
InChI KeyDTCCTIQRPGSLPT-ONEGZZNKSA-N
SMILESCC\C=C\C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4400 to 1.4470 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only