Petroleum ether, for residue analysis, 40-60°C, for trace anal. of polyarom. hydrocarbons
Petroleum ether, for residue analysis, 40-60°C, for trace anal. of polyarom. hydrocarbons
Petroleum ether, for residue analysis, 40-60°C, for trace anal. of polyarom. hydrocarbons
Thermo Scientific Chemicals

Petroleum ether, for residue analysis, 40-60°C, for trace anal. of polyarom. hydrocarbons

CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 326790010
Price (EUR)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS64742-49-0
IUPAC NamePetroleum ether
MDL NumberMFCD00081849
SpecificationsSpecification SheetSpecification Sheet
Water=<0.01 % (Coulometric)
Color scale=<10 APHA
Appearance (Form)Clear liquid
Boiling point40°C to 60°C
Residue after evaporation=<0.0003 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
RUO – Research Use Only