Toluene, 99.85%, Extra Dry, AcroSeal™
Toluene, 99.85%, Extra Dry, AcroSeal™
Toluene, 99.85%, Extra Dry, AcroSeal™
Toluene, 99.85%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

Toluene, 99.85%, Extra Dry, AcroSeal™

CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 326981000
Price (EUR)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.8 %
Water=<0.003 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only