TAPS, 99+%, for biochemistry
TAPS, 99+%, for biochemistry
TAPS, 99+%, for biochemistry
Thermo Scientific Chemicals

TAPS, 99+%, for biochemistry

CAS: 29915-38-6 | C7H17NO6S | 243.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 327800250
Price (EUR)
30,65
Online exclusive
34,30
Save 3,65 (11%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
30,65
Online exclusive
34,30
Save 3,65 (11%)
Each
Chemical Identifiers
CAS29915-38-6
IUPAC Name3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propane-1-sulfonic acid
Molecular FormulaC7H17NO6S
InChI KeyYNLCVAQJIKOXER-UHFFFAOYSA-N
SMILESOCC(CO)(CO)NCCCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH99.0 to 101.0 %
pH5.0 to 6.5 (1 % in water at 25°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only