Methyl trans-2-pentenoate, 95%
Methyl trans-2-pentenoate, 95%
Methyl trans-2-pentenoate, 95%
Thermo Scientific Chemicals

Methyl trans-2-pentenoate, 95%

CAS: 15790-88-2 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Plastic bottle
Catalog number 329720050
Price (EUR)
84,70
Each
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
84,70
Each
Chemical Identifiers
CAS15790-88-2
IUPAC Namemethyl (2E)-pent-2-enoate
Molecular FormulaC6H10O2
InChI KeyMBAHGFJTIVZLFB-SNAWJCMRSA-N
SMILESCC\C=C\C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4300 to 1.4320 (20°C, 589 nm)
GC>=94 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only