N-Hydroxyphthalimide, 98%
N-Hydroxyphthalimide, 98%
N-Hydroxyphthalimide, 98%
Thermo Scientific Chemicals

N-Hydroxyphthalimide, 98%

CAS: 524-38-9 | C8H5NO3 | 163.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic drum
Catalog number 329870025
Price (EUR)
-
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS524-38-9
IUPAC Name2-hydroxy-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC8H5NO3
InChI KeyCFMZSMGAMPBRBE-UHFFFAOYSA-N
SMILESON1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Water=<2 %
Appearance (Color)Yellow
Titration with NaOH97.5 to 102.5 % (On anhydrous substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only