Diethylphosphonoacetic acid, 98%
Diethylphosphonoacetic acid, 98%
Diethylphosphonoacetic acid, 98%
Thermo Scientific Chemicals

Diethylphosphonoacetic acid, 98%

CAS: 3095-95-2 | C6H13O5P | 196.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 330670050
Price (EUR)
31,80
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
31,80
Each
Chemical Identifiers
CAS3095-95-2
IUPAC Name2-(diethoxyphosphoryl)acetate
Molecular FormulaC6H12O5P
InChI KeyDVQMPWOLBFKUMM-UHFFFAOYSA-M
SMILESCCOP(=O)(CC([O-])=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4440 to 1.4460 (20°C, 589 nm)
Titration with NaOH>=97.5 %
Appearance (Color)Clear colorless to yellow
Appearance (Form)Viscous liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only