6-Bromo-2-tetralone, 98%
6-Bromo-2-tetralone, 98%
6-Bromo-2-tetralone, 98%
Thermo Scientific Chemicals

6-Bromo-2-tetralone, 98%

CAS: 4133-35-1 | C10H9BrO | 225.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 330770010
Price (EUR)
51,40
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
51,40
Each
Chemical Identifiers
CAS4133-35-1
IUPAC Name6-bromo-1,2,3,4-tetrahydronaphthalen-2-one
Molecular FormulaC10H9BrO
InChI KeyBYHKDUFPSJWJDI-UHFFFAOYSA-N
SMILESBrC1=CC=C2CC(=O)CCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Light yellow to brown
Melting point70°C to 76°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only