Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)
Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)
Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)
Thermo Scientific Chemicals

Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)

CAS: 9004-32-4 | (C12 H14 O9 R6)n | 263.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 332611000
Price (EUR)
47,65
Online Exclusive
52,30
Save 4,65 (9%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
47,65
Online Exclusive
52,30
Save 4,65 (9%)
Each
Chemical Identifiers
CAS9004-32-4
IUPAC Name2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
Molecular Formula(C12 H14 O9 R6)n
InChI KeyDPXJVFZANSGRMM-UHFFFAOYNA-N
SMILES[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Loss on drying=<10 % (As packed) (3 to 5 g, 105°C, 2 h)
Mercury (Hg)=<1 ppm
Degree of substitution0.65 to 0.90
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only