Polysorbate 40
Polysorbate 40
Polysorbate 40
Thermo Scientific Chemicals

Polysorbate 40

CAS: 9005-66-7 | (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | 522.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 334140025
Price (EUR)
147,00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
147,00
Each
Specifications
CAS9005-66-7
Chemical Name or MaterialPolysorbate 40
ColorYellow
Density1.0500g/mL
Flash Point>200°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only