1,2-Epoxyhexane, 97%
1,2-Epoxyhexane, 97%
1,2-Epoxyhexane, 97%
Thermo Scientific Chemicals

1,2-Epoxyhexane, 97%

CAS: 1436-34-6 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 335551000
Price (EUR)
117,00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
117,00
Each
Chemical Identifiers
CAS1436-34-6
IUPAC Name2-butyloxirane
Molecular FormulaC6H12O
InChI KeyWHNBDXQTMPYBAT-UHFFFAOYNA-N
SMILESCCCCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4040 to 1.4070 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only