n-Hexadecyl palmitate, 98%
n-Hexadecyl palmitate, 98%
n-Hexadecyl palmitate, 98%
Thermo Scientific Chemicals

n-Hexadecyl palmitate, 98%

CAS: 540-10-3 | C32H64O2 | 480.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 338460250
Price (EUR)
72,65
Online exclusive
80,40
Save 7,75 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
72,65
Online exclusive
80,40
Save 7,75 (10%)
Each
Chemical Identifiers
CAS540-10-3
IUPAC Namehexadecyl hexadecanoate
Molecular FormulaC32H64O2
InChI KeyPXDJXZJSCPSGGI-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point52°C to 56°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only