Patent Blue V, sodium salt
Patent Blue V, sodium salt
Patent Blue V, sodium salt
Thermo Scientific Chemicals

Patent Blue V, sodium salt

CAS: 20262-76-4 | C27H31N2NaO7S2 | 582.662 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 339330050
Price (EUR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20262-76-4
IUPAC Namesodium 4-{[4-(diethylamino)phenyl][4-(diethyliminiumyl)cyclohexa-2,5-dien-1-ylidene]methyl}-6-hydroxybenzene-1,3-disulfonate
Molecular FormulaC27H31N2NaO7S2
InChI KeyPMLFOMWMYRKZRF-UHFFFAOYSA-M
SMILES[Na+].CCN(CC)C1=CC=C(C=C1)C(=C1C=CC(C=C1)=[N+](CC)CC)C1=CC(O)=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Dark blue-green
Appearance (Form)Crystalline powder
Dye content>=80 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only