Phenosafranin
Phenosafranin
Phenosafranin
Thermo Scientific Chemicals

Phenosafranin

CAS: 81-93-6 | C18H15ClN4 | 322.80 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3393400505 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 339340050
Price (EUR)
142,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
142,00
Each
Chemical Identifiers
CAS81-93-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Dye content>=80 %
Absorptivity (1%, 1cm)>=1000 (1 % in water, 517-521 nm)
Appearance (Color)Green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only