Sodium perchlorate monohydrate, 85+%, ACS reagent
Sodium perchlorate monohydrate, 85+%, ACS reagent
Sodium perchlorate monohydrate, 85+%, ACS reagent
Sodium perchlorate monohydrate, 85+%, ACS reagent
Thermo Scientific Chemicals

Sodium perchlorate monohydrate, 85+%, ACS reagent

CAS: 7791-07-3 | ClNaO4·H2O | 140.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 343631000
Price (EUR)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7791-07-3
IUPAC Namesodium hydrate perchlorate
Molecular FormulaClH2NaO5
InChI KeyIXGNPUSUVRTQGW-UHFFFAOYSA-M
SMILESO.[Na+].[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH6.0 to 8.0 (5 % solution at 25.0°C)
Insoluble matter=<0.005 %
Chloride (Cl)=<30 ppm
Sulfate (SO4)=<20 ppm
Calcium (Ca)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only