N-Ethylbenzylamine, 95%
N-Ethylbenzylamine, 95%
N-Ethylbenzylamine, 95%
Thermo Scientific Chemicals

N-Ethylbenzylamine, 95%

CAS: 14321-27-8 | C9H13N | 135.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 343715000
Price (EUR)
382,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
382,00
Each
Chemical Identifiers
CAS14321-27-8
IUPAC Namebenzyl(ethyl)amine
Molecular FormulaC9H13N
InChI KeyHVAAHUDGWQAAOJ-UHFFFAOYSA-N
SMILESCCNCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO4>=94 %
Refractive index1.5100 to 1.5130 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only