3-Carboxyphenylboronic acid, 97%
3-Carboxyphenylboronic acid, 97%
3-Carboxyphenylboronic acid, 97%
Thermo Scientific Chemicals

3-Carboxyphenylboronic acid, 97%

CAS: 25487-66-5 | C7H7BO4 | 165.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 344630010
Price (EUR)
48,70
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
48,70
Each
Chemical Identifiers
CAS25487-66-5
IUPAC Name3-(dihydroxyboranyl)benzoic acid
Molecular FormulaC7H7BO4
InChI KeyDBVFWZMQJQMJCB-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only