all-trans-Retinol, 95%
all-trans-Retinol, 95%
all-trans-Retinol, 95%
Thermo Scientific Chemicals

all-trans-Retinol, 95%

All-trans-retinol, Vitamin A, retinol, CAS # 68-26-8, is a fat-soluble vitamin that binds retinoid receptors within cells. | CAS: 68-26-8 | C20H30O | 286.459 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
250 mg
500 mg
1 g
Packaging:
Glass bottle
Catalog number 347070010
Price (EUR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS68-26-8
IUPAC Name(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol
Molecular FormulaC20H30O
InChI KeyFPIPGXGPPPQFEQ-OVSJKPMPSA-N
SMILESC\C(=C/CO)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or powder with lumps
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description


• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient
• All-trans-retinol is involved in hormonal signaling via the retinoid receptors
• It is an important regulator of cell division and apoptosis
• Deficiency can lead to vision, bone, immune, and skin disorders

Applications


• Suitable for research on cell differentiation, embryonic development, immunology and more
• Laboratory grade quality, for research use only

RUO – Research Use Only

General References:

  1. Stephensen, C.B.; Lietz, G. Vitamin A in resistance to and recovery from infection: relevance to SARS-CoV2. Br J Nutr. 2021, 126(11), 1663-1672.
  2. He, J.; Gu, Y.; Zhang, S. Vitamin A and Breast Cancer Survival: A Systematic Review and Meta-analysis. Clin Breast Cancer. 2018, 18(6), e1389-e1400.