Di-μ-hydroxo-bis-[(N,N,N',N'-tetramethylethylenediamine)copper(II)]chloride, 98%
Di-μ-hydroxo-bis-[(N,N,N',N'-tetramethylethylenediamine)copper(II)]chloride, 98%
Di-μ-hydroxo-bis-[(N,N,N',N'-tetramethylethylenediamine)copper(II)]chloride, 98%
Thermo Scientific Chemicals

Di-μ-hydroxo-bis-[(N,N,N',N'-tetramethylethylenediamine)copper(II)]chloride, 98%

CAS: 30698-64-7 | C12H36Cl2Cu2N4O2 | 466.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 348980050
Price (EUR)
55,90
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
55,90
Each
Chemical Identifiers
CAS30698-64-7
IUPAC Namebis([2-(dimethylamino)ethyl]dimethylamine) bis(chlorocopper) dihydrate
Molecular FormulaC12H36Cl2Cu2N4O2
InChI KeyVVXKYYDFGPZSOZ-UHFFFAOYSA-L
SMILESO.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Dark purple
Infrared spectrumConforms
Titration Argentometric>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only