2,3-Diamino-5-bromopyridine, 97%
2,3-Diamino-5-bromopyridine, 97%
2,3-Diamino-5-bromopyridine, 97%
Thermo Scientific Chemicals

2,3-Diamino-5-bromopyridine, 97%

CAS: 38875-53-5 | C5H7BrN3 | 189.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 350330050
Price (EUR)
16,80
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
16,80
Each
Chemical Identifiers
CAS38875-53-5
IUPAC Name2,3-diamino-5-bromopyridin-1-ium
Molecular FormulaC5H7BrN3
InChI KeyYRGMYJUKFJPNPD-UHFFFAOYSA-O
SMILESNC1=CC(Br)=C[NH+]=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or needles
Appearance (Color)Light yellow to light brown to purple
Titration with HClO4>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only