Chloroform-d, for NMR, 99.8 atom % D, stabilized with silver foil
Chloroform-d, for NMR, 99.8 atom % D, stabilized with silver foil
Chloroform-d, for NMR, 99.8 atom % D, stabilized with silver foil
Thermo Scientific Chemicals

Chloroform-d, for NMR, 99.8 atom % D, stabilized with silver foil

CAS: 865-49-6 | CHCl3 | 120.375 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 351420250
Price (EUR)
-
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS865-49-6
IUPAC Nametrichloro(²H)methane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-MICDWDOJSA-N
SMILES[2H]C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.02 % (Coulometric)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
D-Enrichment>=99.75 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only