mono-Methyl terephthalate, 97%
mono-Methyl terephthalate, 97%
mono-Methyl terephthalate, 97%
Thermo Scientific Chemicals

mono-Methyl terephthalate, 97%

CAS: 1679-64-7 | C9H8O4 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 352340050
Price (EUR)
39,30
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
39,30
Each
Chemical Identifiers
CAS1679-64-7
IUPAC Name4-(methoxycarbonyl)benzoic acid
Molecular FormulaC9H8O4
InChI KeyREIDAMBAPLIATC-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=96.0 %
Melting point219°C to 224°C
Appearance (Color)White to off-white
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only