MOPS sodium salt, 98%
MOPS sodium salt, 98%
MOPS sodium salt, 98%
Thermo Scientific Chemicals

MOPS sodium salt, 98%

CAS: 71119-22-7 | C7H14NNaO4S | 231.242 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 352590250
Price (EUR)
63,10
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
63,10
Each
Chemical Identifiers
CAS71119-22-7
IUPAC Namesodium 3-(morpholin-4-yl)propane-1-sulfonate
Molecular FormulaC7H14NNaO4S
InChI KeyMWEMXEWFLIDTSJ-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)CCCN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Solubility(23.125 g/l in cold water) Clear and colorless
Titration with HClO4>=97.5 %
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only