Thermo Scientific Chemicals

Morin hydrate, Thermo Scientific Chemicals

Catalog number: 354000100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Morin hydrate, Thermo Scientific Chemicals

Catalog number: 354000100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 354000100
Price (EUR)
Price: 88,60
Online price: 79,74
Your price:
Quantity
-

Chemical Identifiers

CAS
654055-01-3
IUPAC Name
2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one
Molecular Formula
C15H10O7
InChI Key
YXOLAZRVSSWPPT-UHFFFAOYSA-N
SMILES
OC1=CC(O)=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1
Appearance (Color)
Yellow to brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
UV Lambda max
360 to 375 nm (in methanol)
Specific extinction E (1%/1cm)
>=300 (at Lambda max)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Chronic aquatic toxicity
    Category 2
    Label Elements
    Hazard Statements

    H411 - Toxic to aquatic life with long lasting effects

    Precautionary Statements

    P273 - Avoid release to the environment

    P391 - Collect spillage

    P501 - Dispose of contents/ container to an approved waste disposal plant