TentaGel™ MB-Br
TentaGel™ MB-Br
TentaGel™ MB-Br
Thermo Scientific Chemicals

TentaGel™ MB-Br

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 354480050
Price (EUR)
240,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
240,00
Each
Chemical Identifiers
CAS73342-17-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Loading0.4 to 0.6 mmol/g
Particle size140 to 170 µm
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesis
RUO – Research Use Only