1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis

CAS: 872-50-4 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 354900025
Price (EUR)
344,00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
344,00
Each
Specifications
Assay Percent Range99.4% min. (GC)
Beilstein21,237
Boiling Point202.0°C
CAS872-50-4
Chemical Name or Material1-Methyl-2-pyrrolidinone
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only