3,5-Dimethylbenzyl bromide, 98%
3,5-Dimethylbenzyl bromide, 98%
3,5-Dimethylbenzyl bromide, 98%
Thermo Scientific Chemicals

3,5-Dimethylbenzyl bromide, 98%

CAS: 27129-86-8 | C9H11Br | 199.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 355560050
Price (EUR)
42,90
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
42,90
Each
Chemical Identifiers
CAS27129-86-8
IUPAC Name1-(bromomethyl)-3,5-dimethylbenzene
Molecular FormulaC9H11Br
InChI KeyQXDHXCVJGBTQMK-UHFFFAOYSA-N
SMILESCC1=CC(CBr)=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals or fused mass
GC>=97.5 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
Melting point36°C to 40°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only