2-chloro-6-fluoroaniline, 99%
2-chloro-6-fluoroaniline, 99%
2-chloro-6-fluoroaniline, 99%
Thermo Scientific Chemicals

2-chloro-6-fluoroaniline, 99%

CAS: 363-51-9 | C6H5ClFN | 145.56 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3558200101 gGlass bottle
Catalog number 355820010
Price (EUR)
43,60
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
43,60
Each
Chemical Identifiers
CAS363-51-9
IUPAC Name2-chloro-6-fluoroaniline
Molecular FormulaC6H5ClFN
InChI KeyZJLAWMDJTMMTQB-UHFFFAOYSA-N
SMILESNC1=C(F)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.5480 to 1.5540 (20°C, 589 nm)
Appearance (Color)Clear light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only