n-Nonanoic acid, 97%
n-Nonanoic acid, 97%
n-Nonanoic acid, 97%
Thermo Scientific Chemicals

n-Nonanoic acid, 97%

CAS: 112-05-0 | C9H18O2 | 158.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
View all
Plastic Drum
Catalog number 356240025
Price (EUR)
125,65
Online Exclusive
139,00
Save 13,35 (10%)
Each
Quantity:
2.5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (EUR)
125,65
Online Exclusive
139,00
Save 13,35 (10%)
Each
Chemical Identifiers
CAS112-05-0
IUPAC Namenonanoic acid
Molecular FormulaC9H18O2
InChI KeyFBUKVWPVBMHYJY-UHFFFAOYSA-N
SMILESCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4300 to 1.4340 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only