2-Phenylimidazole, 98%
2-Phenylimidazole, 98%
2-Phenylimidazole, 98%
Thermo Scientific Chemicals

2-Phenylimidazole, 98%

CAS: 670-96-2 | C9H8N2 | 144.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 356431000
Price (EUR)
136,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
136,00
Each
Chemical Identifiers
CAS670-96-2
IUPAC Name2-phenyl-1H-imidazole
Molecular FormulaC9H8N2
InChI KeyZCUJYXPAKHMBAZ-UHFFFAOYSA-N
SMILESN1C=CN=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point142°C to 148°C
Appearance (Color)Light yellow to beige
Appearance (Form)Crystalline or granular powder
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only