Dimethyl 5-hydroxyisophthalate, 99%
Dimethyl 5-hydroxyisophthalate, 99%
Dimethyl 5-hydroxyisophthalate, 99%
Thermo Scientific Chemicals

Dimethyl 5-hydroxyisophthalate, 99%

CAS: 13036-02-7 | C10H10O5 | 210.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 356440100
Price (EUR)
41,70
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
41,70
Each
Chemical Identifiers
CAS13036-02-7
IUPAC Name1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate
Molecular FormulaC10H10O5
InChI KeyDOSDTCPDBPRFHQ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(=CC(O)=C1)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point162°C to 165°C
Appearance (Color)White to light beige
Appearance (Form)Powder and/or lumps
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only