Nickel(II) tetrafluoroborate hexahydrate, 99%
Nickel(II) tetrafluoroborate hexahydrate, 99%
Nickel(II) tetrafluoroborate hexahydrate, 99%
Nickel(II) tetrafluoroborate hexahydrate, 99%
Thermo Scientific Chemicals

Nickel(II) tetrafluoroborate hexahydrate, 99%

CAS: 15684-36-3 | B2F8H12NiO6 | 340.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 358730100
Price (EUR)
32,50
Each
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
32,50
Each
Add to cart
Chemical Identifiers
CAS15684-36-3
IUPAC Namenickel(2+) bis(tetrafluoroboranuide) hexahydrate
Molecular FormulaB2F8H12NiO6
InChI KeyBFFWGFMAXHCLDV-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue-green
Appearance (Form)Powder or crystals
Titration Complexometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only